Gene Transcripts

Gene Symbol
OR52N5
Entrez Gene ID
390075
Gene Name
olfactory receptor family 52 subfamily N member 5
Gene Aliases
OR11-62, OR52N5Q
Chromosome Location
Chr. 11: 5777634 - 5778667 on Build GRCh38
UniGene ID
Hs.690207
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001922.2 NP_001001922.2 1 955
GenBank mRNA BC136979.1 - 1 955
BC136981.1 - 1 955 9020973

View Full Product Details