Gene Transcripts

Gene Symbol
MCCD1
Entrez Gene ID
401250
Gene Name
mitochondrial coiled-coil domain 1
Gene Aliases
Chromosome Location
Chr. 6: 31528962 - 31530231 on Build GRCh38
UniGene ID
Hs.558922
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011700.2 NP_001011700.2 1 94
GenBank mRNA BC132994.1 - 1 76
BC132996.1 - 1 76 40147018

View Full Product Details