Gene Transcripts

Gene Symbol
UBE4B
Entrez Gene ID
10277
Gene Name
ubiquitination factor E4B
Gene Aliases
E4, HDNB1, UBOX3, UFD2, UFD2A
Chromosome Location
Chr. 1: 10032983 - 10181239 on Build GRCh38
UniGene ID
Hs.593974
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105562.2 NP_001099032.1 19, 20 3276
NM_006048.4 NP_006039.2 18, 19 2889
XM_005263422.1 XP_005263479.1 20, 21 3253
XM_011540488.2 XP_011538790.1 19, 20 2552
XM_011540489.2 XP_011538791.1 20, 21 3253
XM_011540490.1 XP_011538792.1 20, 21 3253
XM_017000035.1 XP_016855524.1 17, 18 2339
XM_017000036.1 XP_016855525.1 19, 20 2553
GenBank mRNA AB014584.3 - 18, 19 2780
AB028839.1 - 18, 19 2897
AF043117.1 - 19, 20 2673
AF331520.1 - 18, 19 2248
AK292444.1 - 18, 19 2371
AK304484.1 - 16, 17 1876
BC093696.1 - 18, 19 2279 7939541
BC143458.1 - 18, 19 2279
BC143460.1 - 17, 18 2164
JF289274.1 - 20, 21 2741

View Full Product Details