Gene Transcripts

Gene Symbol
RFC5
Entrez Gene ID
5985
Gene Name
replication factor C subunit 5
Gene Aliases
RFC36
Chromosome Location
Chr. 12: 118016701 - 118033568 on Build GRCh38
UniGene ID
Hs.731908
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017019779.1 XP_016875268.1 9 994

View Full Product Details