Gene Transcripts

Gene Symbol
GYPA
Entrez Gene ID
2993
Gene Name
glycophorin A (MNS blood group)
Gene Aliases
CD235a, GPA, GPErik, GPSAT, HGpMiV, HGpMiXI, HGpSta(C), MN, MNS, PAS-2
Chromosome Location
Chr. 4: 144109303 - 144140854 on Build GRCh38
UniGene ID
Hs.434973
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308190.1 NP_001295119.1 3 299
NM_002099.7 NP_002090.4 4 398
XM_017008134.1 XP_016863623.1 4 402
XM_017008135.1 XP_016863624.1 5 465
XM_017008136.1 XP_016863625.1 3 367
GenBank mRNA AA334250.1 - 4 285
AK290561.1 - 4 363
AY950614.1 - 4 247
BC005319.1 - 4 304 3930700
BC013328.1 - 4 285 4134368
DA537533.1 - 4 363
DA539682.1 - 4 399
EU338231.1 - 5 367
EU338232.1 - 4 284
EU338233.1 - 5 367
EU338234.1 - 3 191
GU347002.1 - 5 391
GU347003.1 - 5 470
GU347004.1 - 5 448
GU347006.1 - 4 290
GU347008.1 - 3 185
L07251.1 - 3 151
L31860.1 - 4 302
M12857.1 - 4 257
M36281.1 - 4 257
U00177.1 - 4 247
U00178.1 - 3 151
U00179.1 - 2 52
X08054.1 - 4 303
X51798.1 - 4 271

View Full Product Details