Gene Transcripts

Gene Symbol
Mfng
Entrez Gene ID
17305
Gene Name
MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Gene Aliases
AW546563
Chromosome Location
Chr. 15: 78755882 - 78773516 on Build GRCm38
UniGene ID
Mm.149235
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008595.2 NP_032621.1 4 638
GenBank mRNA AF015769.1 - 4 465
AK143523.1 - 4 672
BC010983.1 - 4 660 3980692
U94349.1 - 4 638

View Full Product Details