Gene Transcripts

Gene Symbol
Rab22a
Entrez Gene ID
19334
Gene Name
RAB22A, member RAS oncogene family
Gene Aliases
3732413A17Rik, AI662177, AW319644, AW550514, E130120E14Rik, Rab22
Chromosome Location
Chr. 2: 173659845 - 173702182 on Build GRCm38
UniGene ID
Mm.275304
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024436.3 NP_077756.2 3 416
GenBank mRNA AB232616.1 - 3 155
AJ311124.1 - 3 212
AK014407.1 - 402
AK031667.1 - 3 396
AK146594.1 - 3 416
AK149200.1 - 408
AK150617.1 - 3 422
AK219950.1 - 3 161
BC006596.1 - 3 403 3965264

View Full Product Details