Gene Transcripts

Gene Symbol
Ryk
Entrez Gene ID
20187
Gene Name
receptor-like tyrosine kinase
Gene Aliases
AW536699, ERK-3, Vik
Chromosome Location
Chr. 9: 102834920 - 102908307 on Build GRCm38
UniGene ID
Mm.335391
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042607.1 NP_001036072.1 8 1067
NM_001284258.1 NP_001271187.1 7 945
NM_013649.3 NP_038677.3 8 1076
GenBank mRNA AK076449.1 - 2 169
AK144355.1 - 7 885
BC006963.1 - 4 375
BC032275.1 - 8 801 5370100
L02210.1 - 8 1051
L38394.1 - 8 999

View Full Product Details