Gene Transcripts

Gene Symbol
Sftpb
Entrez Gene ID
20388
Gene Name
surfactant associated protein B
Gene Aliases
AI562151, SF-B, SP-B, Sftp-3, Sftp3
Chromosome Location
Chr. 6: 72304610 - 72314373 on Build GRCm38
UniGene ID
Mm.46033
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282071.2 NP_001269000.1 8, 9 937
NM_147779.2 NP_680088.1 8, 9 1006

View Full Product Details