Gene Transcripts

Gene Symbol
Bccip
Entrez Gene ID
66165
Gene Name
BRCA2 and CDKN1A interacting protein
Gene Aliases
1110013J05Rik, TOK-1
Chromosome Location
Chr. 7: 133709333 - 133721145 on Build GRCm38
UniGene ID
Mm.389983
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025392.2 NP_079668.2 2, 3 271
GenBank mRNA AK010684.1 - 2, 3 265
BC048465.1 - 2, 3 263 6534043
CD770748.1 - 2, 3 275

View Full Product Details