Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000429.2 | NP_000420.1 | 7 | 1058 | |
XM_005269842.4 | XP_005269899.1 | 8 | 1288 | ||
XM_005269843.4 | XP_005269900.1 | 7 | 1177 | ||
GenBank mRNA | AK026931.1 | - | 1 | 315 | |
AK290820.1 | - | 7 | 924 | ||
BC018359.1 | - | 7 | 1025 | 4341915 | |
D49357.1 | - | 7 | 821 | ||
X69078.1 | - | 7 | 874 |