Gene Transcripts

Gene Symbol
Mpnd
Entrez Gene ID
68047
Gene Name
MPN domain containing
Gene Aliases
E130307M08Rik
Chromosome Location
Chr. 17: 56009157 - 56016790 on Build GRCm38
UniGene ID
Mm.358710
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026530.5 NP_080806.4 3 525
GenBank mRNA AK012128.1 - 2 480
AK146973.1 - 3 510
AK160356.1 - 3 494
AK172381.1 - 2 268
AK213160.1 - 1 14
BC017625.1 - 2 474

View Full Product Details