Gene Transcripts

Gene Symbol
MKLN1
Entrez Gene ID
4289
Gene Name
muskelin 1
Gene Aliases
TWA2
Chromosome Location
Chr. 7: 131110096 - 131496639 on Build GRCh38
UniGene ID
Hs.44693
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321316.1 NP_001308245.1 1, 2 167
NM_013255.4 NP_037387.2 1, 2 167
XM_006715993.2 XP_006716056.1 1, 2 167
XM_011516224.2 XP_011514526.1 1, 2 167
GenBank mRNA AF047489.1 - 1, 2 121
AK002024.1 - 1, 2 106
AK310452.1 - 1, 2 120
AL705061.1 - 1, 2 111
BC067825.1 - 1, 2 124 5268939
DA172589.1 - 1, 2 167

View Full Product Details