Gene Transcripts

Gene Symbol
Nolc1
Entrez Gene ID
70769
Gene Name
nucleolar and coiled-body phosphoprotein 1
Gene Aliases
NOPP130, NOPP140, P130
Chromosome Location
Chr. 19: 46075847 - 46085543 on Build GRCm38
UniGene ID
Mm.402190
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039351.2 NP_001034440.1 5 744
NM_001039352.2 NP_001034441.1 5 750
NM_001039353.2 NP_001034442.1 5 747
NM_053086.3 NP_444316.2 5 747
GenBank mRNA AK014336.1 - 5 649
AK029150.1 - 5 595
AK034817.1 - 5 624
AK129040.1 - 5 565
AK147118.1 - 5 620
AK150332.1 - 5 581
AK151031.1 - 5 581
AK152941.1 - 5 581
AK166753.1 - 5 653
BC003244.1 - 5 590
CF750495.1 - 5 582

View Full Product Details