Gene Transcripts

Gene Symbol
Stk11ip
Entrez Gene ID
71728
Gene Name
serine/threonine kinase 11 interacting protein
Gene Aliases
1200014D22Rik, BB131189, LIP1, LKB1IP
Chromosome Location
Chr. 1: 75521529 - 75537335 on Build GRCm38
UniGene ID
Mm.312650
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027886.3 NP_082162.3 17 2137
GenBank mRNA AK004757.2 - 17 2090
AK156122.1 - 16 2225
AK171989.1 - 16 1999
AK195907.1 - 2 125
AK213443.1 - 1 43
BC035530.1 - 3 1177
BC042403.1 - 15 2174
BC061635.1 - 17 2120 6530178

View Full Product Details