Gene Transcripts

Gene Symbol
Zfp74
Entrez Gene ID
72723
Gene Name
zinc finger protein 74
Gene Aliases
2810054M15Rik, KRAB8, Zfp66, Znf569, mszf21, mszf77
Chromosome Location
Chr. 7: 29931711 - 29955843 on Build GRCm38
UniGene ID
Mm.371648
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178384.3 NP_848471.2 5 1147
GenBank mRNA BC050048.1 - 5 1147
CF911565.1 - 1 356

View Full Product Details