Gene Transcripts

Gene Symbol
Trp53tg5
Entrez Gene ID
73603
Gene Name
transformation related protein 53 target 5
Gene Aliases
1700126L10Rik
Chromosome Location
Chr. 2: 164467903 - 164478812 on Build GRCm38
UniGene ID
Mm.472883
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271575.1 NP_001258504.1 4 602
GenBank mRNA AK007294.1 - 4 603

View Full Product Details