Gene Transcripts

Gene Symbol
Nrip3
Entrez Gene ID
78593
Gene Name
nuclear receptor interacting protein 3
Gene Aliases
A330103B05Rik, D7H11orf14, ICRFP703B1614Q5.2
Chromosome Location
Chr. 7: 109758056 - 109781545 on Build GRCm38
UniGene ID
Mm.275374
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020610.1 NP_065635.1 4, 5 635
GenBank mRNA AK020731.1 - 4, 5 638
AK083130.1 - 4, 5 637
BC072641.1 - 5, 6 648 6839519

View Full Product Details