Gene Transcripts
- Gene Symbol
- Med8
- Entrez Gene ID
- 80509
- Gene Name
- mediator complex subunit 8
- Gene Aliases
- 2210021A15Rik, AB041805, ARC32
- Chromosome Location
- Chr. 4: 118409298 - 118415824 on Build GRCm38
- UniGene ID
- Mm.259567
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290688.1 |
NP_001277617.1
|
3, 4 |
282 |
|
NM_020000.3 |
NP_064384.2
|
3, 4 |
322 |
|
NM_173719.3 |
NP_776067.1
|
4, 5 |
370 |
|
GenBank mRNA |
AB041805.1 |
- |
2, 3 |
1808 |
|
AK005422.1 |
- |
4, 5 |
353 |
|
AK008759.1 |
- |
3, 4 |
301 |
|
AK011080.1 |
- |
3, 4 |
282 |
|
AK147032.1 |
- |
3, 4 |
308 |
|
AK148025.1 |
- |
3, 4 |
303 |
|
BC002315.1 |
- |
2, 3 |
262 |
|
BC021870.1 |
- |
3, 4 |
293 |
4022630 |
BC051683.1 |
- |
3, 4 |
260 |
|
BC053738.1 |
- |
1, 2 |
1169 |
|
View Full Product Details