Gene Transcripts

Gene Symbol
Pcdhb10
Entrez Gene ID
93881
Gene Name
protocadherin beta 10
Gene Aliases
Pcdhb5D, PcdhbJ
Chromosome Location
Chr. 18: 37411674 - 37414514 on Build GRCm38
UniGene ID
Mm.115996
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053135.2 NP_444365.1 1 1245
GenBank mRNA AY013771.1 - 1 1046
BC112378.1 - 1 1041
BC112397.1 - 1 1045

View Full Product Details