Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36753803-36754300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLCS
Entrez Gene ID
3141
Gene Name
holocarboxylase synthetase
Gene Aliases
HCS
UniGene
Hs.371350
Gene Chromosome Location
Chr.21: 36750888-36990254 on Build GRCh38
Transcripts
NM_000411.6 NM_001242784.1 NM_001242785.1 XM_005260953.3 XM_005260955.3 XM_005260956.3 XM_006723994.2 XM_006723995.1 XM_011529538.1 XM_011529539.2 XM_011529541.2 XM_017028330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5936666 c.2138C>T p.S713F Chr.21: 36754289-36754289 on Build GRCh38
7256469 c.2171A>C p.K724T Chr.21: 36754256-36754256 on Build GRCh38
1414011 c.2146A>T p.M716L Chr.21: 36754281-36754281 on Build GRCh38
5554323 c.2142C>T p.F714F Chr.21: 36754285-36754285 on Build GRCh38
4690680 c.2127G>A p.P709P Chr.21: 36754300-36754300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs137949852 Chr.21: 36754227-36754227 on Build GRCh38
rs750750219 Chr.21: 36754205-36754205 on Build GRCh38
rs957728782 Chr.21: 36753920-36753920 on Build GRCh38
rs73398122 Chr.21: 36753987-36753987 on Build GRCh38
rs1025223317 Chr.21: 36754158-36754158 on Build GRCh38
rs527369571 Chr.21: 36753957-36753957 on Build GRCh38
rs925025089 Chr.21: 36753913-36753913 on Build GRCh38
rs966883214 Chr.21: 36754159-36754159 on Build GRCh38
rs76983917 Chr.21: 36753974-36753974 on Build GRCh38
rs746538591 Chr.21: 36754242-36754242 on Build GRCh38
rs551333782 Chr.21: 36753893-36753893 on Build GRCh38
rs565016948 Chr.21: 36753940-36753940 on Build GRCh38
rs946894707 Chr.21: 36754070-36754070 on Build GRCh38
rs767521131 Chr.21: 36754216-36754216 on Build GRCh38
rs191880316 Chr.21: 36753871-36753871 on Build GRCh38
rs778616712 Chr.21: 36754232-36754232 on Build GRCh38
rs371819968 Chr.21: 36753995-36753995 on Build GRCh38
rs745584892 Chr.21: 36754234-36754234 on Build GRCh38
rs372906843 Chr.21: 36754224-36754224 on Build GRCh38
rs767338333 Chr.21: 36754270-36754271 on Build GRCh38
rs914170919 Chr.21: 36754049-36754049 on Build GRCh38
rs886057074 Chr.21: 36753975-36753976 on Build GRCh38
rs890088284 Chr.21: 36754186-36754186 on Build GRCh38
rs372467001 Chr.21: 36754267-36754267 on Build GRCh38
rs765675311 Chr.21: 36754204-36754204 on Build GRCh38
rs966436737 Chr.21: 36753903-36753903 on Build GRCh38
rs988330603 Chr.21: 36754034-36754034 on Build GRCh38
rs771853376 Chr.21: 36754238-36754238 on Build GRCh38
rs531240761 Chr.21: 36753877-36753877 on Build GRCh38
rs762341755 Chr.21: 36754203-36754203 on Build GRCh38
rs777607443 Chr.21: 36754225-36754225 on Build GRCh38
rs774356408 Chr.21: 36754254-36754257 on Build GRCh38
rs1014853378 Chr.21: 36753845-36753845 on Build GRCh38
rs530027358 Chr.21: 36753999-36753999 on Build GRCh38
rs773627696 Chr.21: 36754250-36754250 on Build GRCh38
rs940499246 Chr.21: 36754035-36754035 on Build GRCh38
rs980034869 Chr.21: 36754125-36754125 on Build GRCh38
rs745974918 Chr.21: 36754228-36754228 on Build GRCh38
rs759489833 Chr.21: 36753946-36753946 on Build GRCh38
rs538687087 Chr.21: 36754090-36754090 on Build GRCh38
rs375809125 Chr.21: 36754246-36754246 on Build GRCh38
rs118099059 Chr.21: 36754078-36754078 on Build GRCh38
rs774549376 Chr.21: 36754258-36754258 on Build GRCh38
rs995090852 Chr.21: 36753885-36753885 on Build GRCh38
rs921887441 Chr.21: 36753974-36753974 on Build GRCh38
rs201681436 Chr.21: 36754237-36754239 on Build GRCh38
rs999634482 Chr.21: 36754166-36754166 on Build GRCh38
rs77014096 Chr.21: 36754206-36754206 on Build GRCh38
rs761447836 Chr.21: 36754268-36754268 on Build GRCh38
rs749078688 Chr.21: 36754228-36754228 on Build GRCh38
rs759699224 Chr.21: 36754262-36754262 on Build GRCh38
rs759245601 Chr.21: 36754136-36754136 on Build GRCh38
rs199860744 Chr.21: 36754213-36754213 on Build GRCh38
rs558525705 Chr.21: 36754165-36754165 on Build GRCh38
rs368947121 Chr.21: 36754252-36754252 on Build GRCh38
rs977851542 Chr.21: 36753912-36753912 on Build GRCh38
rs777271467 Chr.21: 36754200-36754200 on Build GRCh38
rs1050181603 Chr.21: 36754171-36754171 on Build GRCh38
rs1027725702 Chr.21: 36753886-36753886 on Build GRCh38
rs980791735 Chr.21: 36753973-36753973 on Build GRCh38
rs762386073 Chr.21: 36754226-36754226 on Build GRCh38
rs895408176 Chr.21: 36754142-36754142 on Build GRCh38
rs767752067 Chr.21: 36754263-36754263 on Build GRCh38
rs1014216104 Chr.21: 36754154-36754154 on Build GRCh38
rs147474255 Chr.21: 36754253-36754253 on Build GRCh38
rs1037148759 Chr.21: 36754054-36754054 on Build GRCh38
rs554746203 Chr.21: 36754231-36754231 on Build GRCh38
rs775080501 Chr.21: 36754241-36754241 on Build GRCh38
rs182629388 Chr.21: 36754081-36754081 on Build GRCh38
rs199499263 Chr.21: 36754254-36754254 on Build GRCh38
rs763666612 Chr.21: 36754232-36754232 on Build GRCh38
rs886057073 Chr.21: 36753912-36753912 on Build GRCh38
rs995527852 Chr.21: 36754080-36754080 on Build GRCh38
rs969900894 Chr.21: 36753928-36753928 on Build GRCh38
rs898565327 Chr.21: 36754055-36754055 on Build GRCh38
rs931759194 Chr.21: 36754155-36754155 on Build GRCh38
rs763351170 Chr.21: 36754251-36754251 on Build GRCh38
rs974361872 Chr.21: 36753968-36753968 on Build GRCh38
rs567514586 Chr.21: 36753930-36753930 on Build GRCh38
rs766589766 Chr.21: 36754212-36754212 on Build GRCh38
rs770504850 Chr.21: 36753880-36753880 on Build GRCh38

View Full Product Details