Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36759648-36760159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLCS
Entrez Gene ID
3141
Gene Name
holocarboxylase synthetase
Gene Aliases
HCS
UniGene
Hs.371350
Gene Chromosome Location
Chr.21: 36750888-36990254 on Build GRCh38
Transcripts
NM_000411.6 NM_001242784.1 NM_001242785.1 XM_005260953.3 XM_005260955.3 XM_005260956.3 XM_006723994.2 XM_006723995.1 XM_011529538.1 XM_011529539.2 XM_011529541.2 XM_017028330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6018062 c.1681-6C>T p.? Chr.21: 36759847-36759847 on Build GRCh38
2844758 c.1723A>G p.S575G Chr.21: 36759799-36759799 on Build GRCh38
123615 c.1688A>G p.N563S Chr.21: 36759834-36759834 on Build GRCh38
1209782 c.1711G>A p.D571N Chr.21: 36759811-36759811 on Build GRCh38
2844757 c.1740C>T p.I580I Chr.21: 36759782-36759782 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982415251 Chr.21: 36759770-36759770 on Build GRCh38
rs143892712 Chr.21: 36759747-36759747 on Build GRCh38
rs1006065168 Chr.21: 36759977-36759977 on Build GRCh38
rs201167553 Chr.21: 36759733-36759733 on Build GRCh38
rs776257581 Chr.21: 36759714-36759714 on Build GRCh38
rs1025897854 Chr.21: 36759968-36759968 on Build GRCh38
rs754860873 Chr.21: 36759874-36759874 on Build GRCh38
rs543256742 Chr.21: 36759748-36759748 on Build GRCh38
rs769446135 Chr.21: 36759828-36759828 on Build GRCh38
rs369204579 Chr.21: 36759994-36759994 on Build GRCh38
rs762128307 Chr.21: 36759859-36759859 on Build GRCh38
rs764724435 Chr.21: 36759847-36759847 on Build GRCh38
rs554677294 Chr.21: 36759699-36759699 on Build GRCh38
rs761461715 Chr.21: 36759729-36759729 on Build GRCh38
rs74993434 Chr.21: 36759948-36759948 on Build GRCh38
rs764487201 Chr.21: 36759755-36759755 on Build GRCh38
rs966747966 Chr.21: 36760105-36760105 on Build GRCh38
rs984675997 Chr.21: 36759796-36759796 on Build GRCh38
rs759595530 Chr.21: 36759872-36759872 on Build GRCh38
rs889933800 Chr.21: 36759917-36759917 on Build GRCh38
rs752143567 Chr.21: 36760069-36760069 on Build GRCh38
rs119103228 Chr.21: 36759811-36759811 on Build GRCh38
rs753316170 Chr.21: 36759753-36759753 on Build GRCh38
rs148845011 Chr.21: 36759876-36759876 on Build GRCh38
rs562250576 Chr.21: 36759961-36759961 on Build GRCh38
rs1034040858 Chr.21: 36759999-36759999 on Build GRCh38
rs989745706 Chr.21: 36760104-36760104 on Build GRCh38
rs970726967 Chr.21: 36760002-36760002 on Build GRCh38
rs918020522 Chr.21: 36759856-36759856 on Build GRCh38
rs773144232 Chr.21: 36760113-36760113 on Build GRCh38
rs1019121292 Chr.21: 36759710-36759710 on Build GRCh38
rs887462241 Chr.21: 36759971-36759971 on Build GRCh38
rs2073420 Chr.21: 36759871-36759871 on Build GRCh38
rs745960696 Chr.21: 36759682-36759682 on Build GRCh38
rs747012912 Chr.21: 36759804-36759804 on Build GRCh38
rs119103230 Chr.21: 36759781-36759781 on Build GRCh38
rs774004352 Chr.21: 36759835-36759835 on Build GRCh38
rs147273535 Chr.21: 36759751-36759751 on Build GRCh38
rs944234488 Chr.21: 36759921-36759921 on Build GRCh38
rs748819971 Chr.21: 36759833-36759833 on Build GRCh38
rs779949238 Chr.21: 36759802-36759802 on Build GRCh38
rs758454457 Chr.21: 36759794-36759794 on Build GRCh38
rs530868173 Chr.21: 36760057-36760057 on Build GRCh38
rs959216023 Chr.21: 36760020-36760020 on Build GRCh38
rs747942970 Chr.21: 36759888-36759888 on Build GRCh38
rs894312140 Chr.21: 36759941-36759941 on Build GRCh38
rs768494417 Chr.21: 36759707-36759707 on Build GRCh38
rs554690993 Chr.21: 36760021-36760021 on Build GRCh38
rs1025048515 Chr.21: 36760101-36760101 on Build GRCh38
rs188336563 Chr.21: 36759909-36759909 on Build GRCh38
rs747916823 Chr.21: 36759814-36759814 on Build GRCh38
rs754271379 Chr.21: 36759769-36759769 on Build GRCh38
rs148709879 Chr.21: 36759812-36759812 on Build GRCh38
rs375456567 Chr.21: 36759782-36759782 on Build GRCh38
rs376899782 Chr.21: 36759778-36759778 on Build GRCh38
rs374957099 Chr.21: 36759678-36759678 on Build GRCh38
rs1031265752 Chr.21: 36760024-36760024 on Build GRCh38
rs772791252 Chr.21: 36759829-36759829 on Build GRCh38
rs750666707 Chr.21: 36759866-36759866 on Build GRCh38
rs765472058 Chr.21: 36759865-36759865 on Build GRCh38
rs370277326 Chr.21: 36759779-36759779 on Build GRCh38
rs769344210 Chr.21: 36759734-36759734 on Build GRCh38
rs761117077 Chr.21: 36759754-36759754 on Build GRCh38
rs777016577 Chr.21: 36759857-36759857 on Build GRCh38
rs978176095 Chr.21: 36760111-36760111 on Build GRCh38
rs993116401 Chr.21: 36759963-36759963 on Build GRCh38
rs1012321402 Chr.21: 36759947-36759947 on Build GRCh38
rs957143697 Chr.21: 36760091-36760093 on Build GRCh38
rs770632911 Chr.21: 36759834-36759834 on Build GRCh38
rs926435968 Chr.21: 36759798-36759798 on Build GRCh38
rs936870255 Chr.21: 36759895-36759895 on Build GRCh38
rs761209355 Chr.21: 36759839-36759839 on Build GRCh38
rs574447499 Chr.21: 36759704-36759704 on Build GRCh38
rs780004309 Chr.21: 36759701-36759701 on Build GRCh38

View Full Product Details