Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36756118-36756717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLCS
Entrez Gene ID
3141
Gene Name
holocarboxylase synthetase
Gene Aliases
HCS
UniGene
Hs.371350
Gene Chromosome Location
Chr.21: 36750888-36990254 on Build GRCh38
Transcripts
NM_000411.6 NM_001242784.1 NM_001242785.1 XM_005260953.3 XM_005260955.3 XM_005260956.3 XM_006723994.2 XM_006723995.1 XM_011529538.1 XM_011529539.2 XM_011529541.2 XM_017028330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2844755 c.1922T>C p.V641A Chr.21: 36756629-36756629 on Build GRCh38
4101489 c.1911C>A p.I637I Chr.21: 36756640-36756640 on Build GRCh38
4101488 c.1921G>C p.V641L Chr.21: 36756630-36756630 on Build GRCh38
5923383 c.1900G>A p.D634N Chr.21: 36756651-36756651 on Build GRCh38
4930530 c.1965G>T p.G655G Chr.21: 36756586-36756586 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867914308 Chr.21: 36756310-36756310 on Build GRCh38
rs373944714 Chr.21: 36756535-36756535 on Build GRCh38
rs942047515 Chr.21: 36756278-36756278 on Build GRCh38
rs150263699 Chr.21: 36756640-36756640 on Build GRCh38
rs941172739 Chr.21: 36756385-36756385 on Build GRCh38
rs964874998 Chr.21: 36756517-36756517 on Build GRCh38
rs79961519 Chr.21: 36756221-36756221 on Build GRCh38
rs558205337 Chr.21: 36756364-36756364 on Build GRCh38
rs984129285 Chr.21: 36756186-36756186 on Build GRCh38
rs756948819 Chr.21: 36756283-36756283 on Build GRCh38
rs563522883 Chr.21: 36756160-36756160 on Build GRCh38
rs57508268 Chr.21: 36756214-36756214 on Build GRCh38
rs181325364 Chr.21: 36756205-36756205 on Build GRCh38
rs867907621 Chr.21: 36756376-36756376 on Build GRCh38
rs558119330 Chr.21: 36756617-36756617 on Build GRCh38
rs755670854 Chr.21: 36756507-36756507 on Build GRCh38
rs773979089 Chr.21: 36756573-36756573 on Build GRCh38
rs530955703 Chr.21: 36756161-36756161 on Build GRCh38
rs570222602 Chr.21: 36756235-36756235 on Build GRCh38
rs747302103 Chr.21: 36756607-36756607 on Build GRCh38
rs1021667517 Chr.21: 36756240-36756240 on Build GRCh38
rs146448211 Chr.21: 36756558-36756558 on Build GRCh38
rs533994333 Chr.21: 36756282-36756283 on Build GRCh38
rs776535574 Chr.21: 36756560-36756560 on Build GRCh38
rs200453837 Chr.21: 36756647-36756647 on Build GRCh38
rs369812223 Chr.21: 36756154-36756154 on Build GRCh38
rs763530346 Chr.21: 36756583-36756583 on Build GRCh38
rs760372711 Chr.21: 36756630-36756631 on Build GRCh38
rs963823957 Chr.21: 36756367-36756367 on Build GRCh38
rs556965051 Chr.21: 36756254-36756254 on Build GRCh38
rs572665044 Chr.21: 36756243-36756243 on Build GRCh38
rs749197530 Chr.21: 36756280-36756280 on Build GRCh38
rs569435778 Chr.21: 36756225-36756225 on Build GRCh38
rs186604557 Chr.21: 36756267-36756267 on Build GRCh38
rs758017719 Chr.21: 36756452-36756455 on Build GRCh38
rs908452359 Chr.21: 36756570-36756570 on Build GRCh38
rs570083821 Chr.21: 36756272-36756272 on Build GRCh38
rs35509622 Chr.21: 36756288-36756289 on Build GRCh38
rs975169430 Chr.21: 36756368-36756368 on Build GRCh38
rs150665851 Chr.21: 36756630-36756630 on Build GRCh38
rs559185308 Chr.21: 36756451-36756460 on Build GRCh38
rs998049984 Chr.21: 36756174-36756174 on Build GRCh38
rs532791872 Chr.21: 36756446-36756446 on Build GRCh38
rs9977769 Chr.21: 36756406-36756406 on Build GRCh38
rs917374658 Chr.21: 36756157-36756157 on Build GRCh38
rs971659953 Chr.21: 36756247-36756247 on Build GRCh38
rs762915830 Chr.21: 36756568-36756568 on Build GRCh38
rs866644112 Chr.21: 36756624-36756624 on Build GRCh38
rs868172527 Chr.21: 36756311-36756311 on Build GRCh38
rs893453553 Chr.21: 36756229-36756229 on Build GRCh38
rs780639790 Chr.21: 36756590-36756590 on Build GRCh38
rs747420311 Chr.21: 36756538-36756538 on Build GRCh38
rs148896807 Chr.21: 36756177-36756177 on Build GRCh38
rs1025710756 Chr.21: 36756176-36756176 on Build GRCh38
rs183302013 Chr.21: 36756662-36756662 on Build GRCh38
rs575507555 Chr.21: 36756360-36756360 on Build GRCh38
rs61732507 Chr.21: 36756631-36756631 on Build GRCh38
rs748447457 Chr.21: 36756621-36756621 on Build GRCh38
rs565221424 Chr.21: 36756445-36756445 on Build GRCh38
rs140951243 Chr.21: 36756564-36756564 on Build GRCh38
rs544173760 Chr.21: 36756438-36756438 on Build GRCh38
rs191119664 Chr.21: 36756520-36756520 on Build GRCh38
rs746393280 Chr.21: 36756455-36756456 on Build GRCh38
rs149748482 Chr.21: 36756166-36756166 on Build GRCh38
rs756354473 Chr.21: 36756689-36756689 on Build GRCh38
rs1053162614 Chr.21: 36756642-36756642 on Build GRCh38
rs566206357 Chr.21: 36756307-36756307 on Build GRCh38
rs140925558 Chr.21: 36756286-36756286 on Build GRCh38
rs144116568 Chr.21: 36756549-36756549 on Build GRCh38
rs561953171 Chr.21: 36756193-36756193 on Build GRCh38
rs748535375 Chr.21: 36756681-36756681 on Build GRCh38
rs549306707 Chr.21: 36756265-36756265 on Build GRCh38
rs755415494 Chr.21: 36756609-36756609 on Build GRCh38
rs878852989 Chr.21: 36756546-36756546 on Build GRCh38
rs760265551 Chr.21: 36756652-36756652 on Build GRCh38
rs370339949 Chr.21: 36756407-36756407 on Build GRCh38
rs551755928 Chr.21: 36756451-36756454 on Build GRCh38
rs766430268 Chr.21: 36756571-36756571 on Build GRCh38
rs555264391 Chr.21: 36756343-36756343 on Build GRCh38
rs574039619 Chr.21: 36756405-36756405 on Build GRCh38
rs373807983 Chr.21: 36756400-36756400 on Build GRCh38
rs533458583 Chr.21: 36756209-36756209 on Build GRCh38
rs538159086 Chr.21: 36756585-36756585 on Build GRCh38
rs866969316 Chr.21: 36756264-36756264 on Build GRCh38
rs781287179 Chr.21: 36756258-36756258 on Build GRCh38
rs199517800 Chr.21: 36756660-36756660 on Build GRCh38
rs536116974 Chr.21: 36756155-36756155 on Build GRCh38
rs567390172 Chr.21: 36756181-36756181 on Build GRCh38
rs578046388 Chr.21: 36756383-36756383 on Build GRCh38
rs749413115 Chr.21: 36756632-36756632 on Build GRCh38
rs1021814864 Chr.21: 36756357-36756357 on Build GRCh38
rs373220213 Chr.21: 36756274-36756275 on Build GRCh38
rs993514214 Chr.21: 36756203-36756203 on Build GRCh38
rs759023905 Chr.21: 36756248-36756248 on Build GRCh38
rs182012918 Chr.21: 36756249-36756249 on Build GRCh38
rs755620019 Chr.21: 36756579-36756579 on Build GRCh38
rs189379984 Chr.21: 36756234-36756234 on Build GRCh38
rs371198062 Chr.21: 36756271-36756271 on Build GRCh38
rs1024493523 Chr.21: 36756246-36756246 on Build GRCh38
rs34389255 Chr.21: 36756237-36756238 on Build GRCh38
rs558424313 Chr.21: 36756295-36756295 on Build GRCh38
rs942356922 Chr.21: 36756231-36756231 on Build GRCh38
rs138952612 Chr.21: 36756643-36756643 on Build GRCh38
rs1038205736 Chr.21: 36756390-36756390 on Build GRCh38
rs77945621 Chr.21: 36756220-36756220 on Build GRCh38
rs547228125 Chr.21: 36756213-36756213 on Build GRCh38
rs942327563 Chr.21: 36756253-36756253 on Build GRCh38
rs951217851 Chr.21: 36756414-36756414 on Build GRCh38
rs113722511 Chr.21: 36756216-36756216 on Build GRCh38
rs796051978 Chr.21: 36756660-36756660 on Build GRCh38
rs551620897 Chr.21: 36756436-36756436 on Build GRCh38
rs1018223338 Chr.21: 36756168-36756168 on Build GRCh38
rs747473051 Chr.21: 36756451-36756455 on Build GRCh38
rs948770929 Chr.21: 36756402-36756402 on Build GRCh38
rs370117126 Chr.21: 36756577-36756577 on Build GRCh38
rs763760775 Chr.21: 36756653-36756653 on Build GRCh38
rs188089721 Chr.21: 36756146-36756146 on Build GRCh38
rs560684594 Chr.21: 36756261-36756261 on Build GRCh38
rs530968102 Chr.21: 36756195-36756195 on Build GRCh38
rs56071863 Chr.21: 36756475-36756478 on Build GRCh38
rs772576193 Chr.21: 36756527-36756527 on Build GRCh38
rs759416272 Chr.21: 36756574-36756574 on Build GRCh38
rs552767274 Chr.21: 36756242-36756242 on Build GRCh38
rs991675718 Chr.21: 36756201-36756201 on Build GRCh38
rs147517312 Chr.21: 36756663-36756663 on Build GRCh38
rs938898566 Chr.21: 36756287-36756287 on Build GRCh38
rs145668464 Chr.21: 36756171-36756171 on Build GRCh38
rs971375283 Chr.21: 36756303-36756303 on Build GRCh38
rs765032759 Chr.21: 36756283-36756284 on Build GRCh38
rs58201045 Chr.21: 36756429-36756429 on Build GRCh38
rs998017463 Chr.21: 36756238-36756238 on Build GRCh38
rs572801897 Chr.21: 36756187-36756187 on Build GRCh38
rs544260888 Chr.21: 36756204-36756204 on Build GRCh38
rs537706097 Chr.21: 36756361-36756361 on Build GRCh38
rs539098878 Chr.21: 36756241-36756241 on Build GRCh38
rs778182226 Chr.21: 36756692-36756692 on Build GRCh38
rs752162795 Chr.21: 36756666-36756666 on Build GRCh38
rs915024923 Chr.21: 36756252-36756252 on Build GRCh38
rs879079795 Chr.21: 36756451-36756466 on Build GRCh38
rs554271449 Chr.21: 36756156-36756156 on Build GRCh38
rs779383019 Chr.21: 36756511-36756511 on Build GRCh38
rs1006774316 Chr.21: 36756167-36756167 on Build GRCh38
rs147621612 Chr.21: 36756164-36756164 on Build GRCh38
rs755293772 Chr.21: 36756673-36756673 on Build GRCh38
rs368124997 Chr.21: 36756576-36756576 on Build GRCh38
rs367725468 Chr.21: 36756646-36756646 on Build GRCh38
rs370093636 Chr.21: 36756324-36756324 on Build GRCh38
rs540617151 Chr.21: 36756387-36756387 on Build GRCh38
rs1047390663 Chr.21: 36756382-36756382 on Build GRCh38
rs186525725 Chr.21: 36756224-36756224 on Build GRCh38
rs893430655 Chr.21: 36756430-36756430 on Build GRCh38
rs567688437 Chr.21: 36756478-36756478 on Build GRCh38
rs187818323 Chr.21: 36756266-36756266 on Build GRCh38
rs983948893 Chr.21: 36756422-36756423 on Build GRCh38
rs9636905 Chr.21: 36756255-36756255 on Build GRCh38
rs201069938 Chr.21: 36756563-36756563 on Build GRCh38
rs144598736 Chr.21: 36756618-36756618 on Build GRCh38
rs758851021 Chr.21: 36756586-36756586 on Build GRCh38
rs923709958 Chr.21: 36756210-36756210 on Build GRCh38
rs1047374251 Chr.21: 36756239-36756239 on Build GRCh38
rs749358137 Chr.21: 36756694-36756694 on Build GRCh38
rs958699648 Chr.21: 36756197-36756197 on Build GRCh38
rs989606932 Chr.21: 36756251-36756251 on Build GRCh38
rs529526420 Chr.21: 36756263-36756263 on Build GRCh38
rs149399432 Chr.21: 36756651-36756651 on Build GRCh38
rs781552946 Chr.21: 36756678-36756678 on Build GRCh38
rs375847542 Chr.21: 36756245-36756245 on Build GRCh38
rs957580309 Chr.21: 36756250-36756250 on Build GRCh38
rs752117681 Chr.21: 36756491-36756491 on Build GRCh38
rs386818320 Chr.21: 36756266-36756267 on Build GRCh38
rs58695786 Chr.21: 36756254-36756255 on Build GRCh38
rs541641199 Chr.21: 36756244-36756244 on Build GRCh38
rs201679041 Chr.21: 36756479-36756479 on Build GRCh38
rs374881871 Chr.21: 36756158-36756158 on Build GRCh38
rs546186992 Chr.21: 36756449-36756449 on Build GRCh38
rs922222257 Chr.21: 36756371-36756371 on Build GRCh38
rs557047143 Chr.21: 36756165-36756165 on Build GRCh38
rs145280272 Chr.21: 36756188-36756188 on Build GRCh38
rs976589227 Chr.21: 36756262-36756262 on Build GRCh38
rs540702554 Chr.21: 36756257-36756257 on Build GRCh38

View Full Product Details