Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36754135-36754388 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLCS
Entrez Gene ID
3141
Gene Name
holocarboxylase synthetase
Gene Aliases
HCS
UniGene
Hs.371350
Gene Chromosome Location
Chr.21: 36750888-36990254 on Build GRCh38
Transcripts
NM_000411.6 NM_001242784.1 NM_001242785.1 XM_005260953.3 XM_005260955.3 XM_005260956.3 XM_006723994.2 XM_006723995.1 XM_011529538.1 XM_011529539.2 XM_011529541.2 XM_017028330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5559913 c.2085C>G p.L695L Chr.21: 36754342-36754342 on Build GRCh38
5936666 c.2138C>T p.S713F Chr.21: 36754289-36754289 on Build GRCh38
5556706 c.2092C>T p.H698Y Chr.21: 36754335-36754335 on Build GRCh38
7256469 c.2171A>C p.K724T Chr.21: 36754256-36754256 on Build GRCh38
1414011 c.2146A>T p.M716L Chr.21: 36754281-36754281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs137949852 Chr.21: 36754227-36754227 on Build GRCh38
rs750750219 Chr.21: 36754205-36754205 on Build GRCh38
rs371982620 Chr.21: 36754296-36754296 on Build GRCh38
rs774651759 Chr.21: 36754338-36754338 on Build GRCh38
rs768097284 Chr.21: 36754321-36754321 on Build GRCh38
rs752737867 Chr.21: 36754283-36754283 on Build GRCh38
rs746538591 Chr.21: 36754242-36754242 on Build GRCh38
rs202028605 Chr.21: 36754297-36754297 on Build GRCh38
rs767521131 Chr.21: 36754216-36754216 on Build GRCh38
rs745584892 Chr.21: 36754234-36754234 on Build GRCh38
rs778616712 Chr.21: 36754232-36754232 on Build GRCh38
rs145337682 Chr.21: 36754320-36754320 on Build GRCh38
rs372906843 Chr.21: 36754224-36754224 on Build GRCh38
rs767338333 Chr.21: 36754270-36754271 on Build GRCh38
rs890088284 Chr.21: 36754186-36754186 on Build GRCh38
rs372467001 Chr.21: 36754267-36754267 on Build GRCh38
rs765675311 Chr.21: 36754204-36754204 on Build GRCh38
rs778781151 Chr.21: 36754298-36754298 on Build GRCh38
rs771853376 Chr.21: 36754238-36754238 on Build GRCh38
rs532108774 Chr.21: 36754285-36754285 on Build GRCh38
rs762341755 Chr.21: 36754203-36754203 on Build GRCh38
rs777607443 Chr.21: 36754225-36754225 on Build GRCh38
rs774356408 Chr.21: 36754254-36754257 on Build GRCh38
rs140014588 Chr.21: 36754284-36754284 on Build GRCh38
rs199936080 Chr.21: 36754363-36754363 on Build GRCh38
rs758074977 Chr.21: 36754304-36754304 on Build GRCh38
rs773627696 Chr.21: 36754250-36754250 on Build GRCh38
rs761596791 Chr.21: 36754357-36754357 on Build GRCh38
rs767700692 Chr.21: 36754339-36754339 on Build GRCh38
rs773752745 Chr.21: 36754323-36754323 on Build GRCh38
rs745974918 Chr.21: 36754228-36754228 on Build GRCh38
rs145648338 Chr.21: 36754368-36754368 on Build GRCh38
rs375809125 Chr.21: 36754246-36754246 on Build GRCh38
rs774549376 Chr.21: 36754258-36754258 on Build GRCh38
rs201681436 Chr.21: 36754237-36754239 on Build GRCh38
rs999634482 Chr.21: 36754166-36754166 on Build GRCh38
rs77014096 Chr.21: 36754206-36754206 on Build GRCh38
rs761447836 Chr.21: 36754268-36754268 on Build GRCh38
rs749078688 Chr.21: 36754228-36754228 on Build GRCh38
rs759699224 Chr.21: 36754262-36754262 on Build GRCh38
rs750759085 Chr.21: 36754303-36754303 on Build GRCh38
rs199860744 Chr.21: 36754213-36754213 on Build GRCh38
rs368947121 Chr.21: 36754252-36754252 on Build GRCh38
rs149733264 Chr.21: 36754358-36754358 on Build GRCh38
rs142437842 Chr.21: 36754324-36754324 on Build GRCh38
rs777271467 Chr.21: 36754200-36754200 on Build GRCh38
rs773191073 Chr.21: 36754364-36754365 on Build GRCh38
rs1050181603 Chr.21: 36754171-36754171 on Build GRCh38
rs762386073 Chr.21: 36754226-36754226 on Build GRCh38
rs79373682 Chr.21: 36754331-36754331 on Build GRCh38
rs779883958 Chr.21: 36754312-36754312 on Build GRCh38
rs767752067 Chr.21: 36754263-36754263 on Build GRCh38
rs147474255 Chr.21: 36754253-36754253 on Build GRCh38
rs554746203 Chr.21: 36754231-36754231 on Build GRCh38
rs775080501 Chr.21: 36754241-36754241 on Build GRCh38
rs779826873 Chr.21: 36754369-36754369 on Build GRCh38
rs764148793 Chr.21: 36754349-36754349 on Build GRCh38
rs199499263 Chr.21: 36754254-36754254 on Build GRCh38
rs763666612 Chr.21: 36754232-36754232 on Build GRCh38
rs745670538 Chr.21: 36754300-36754300 on Build GRCh38
rs147256659 Chr.21: 36754342-36754342 on Build GRCh38
rs763351170 Chr.21: 36754251-36754251 on Build GRCh38
rs766589766 Chr.21: 36754212-36754212 on Build GRCh38
rs139697458 Chr.21: 36754356-36754356 on Build GRCh38

View Full Product Details