Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36754335-36754582 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLCS
Entrez Gene ID
3141
Gene Name
holocarboxylase synthetase
Gene Aliases
HCS
UniGene
Hs.371350
Gene Chromosome Location
Chr.21: 36750888-36990254 on Build GRCh38
Transcripts
NM_000411.6 NM_001242784.1 NM_001242785.1 XM_005260953.3 XM_005260955.3 XM_005260956.3 XM_006723994.2 XM_006723995.1 XM_011529538.1 XM_011529539.2 XM_011529541.2 XM_017028330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5559913 c.2085C>G p.L695L Chr.21: 36754342-36754342 on Build GRCh38
5556706 c.2092C>T p.H698Y Chr.21: 36754335-36754335 on Build GRCh38
3550570 c.2023C>T p.H675Y Chr.21: 36754404-36754404 on Build GRCh38
2844752 c.2056A>G p.I686V Chr.21: 36754371-36754371 on Build GRCh38
6251633 c.2055C>A p.S685S Chr.21: 36754372-36754372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368743397 Chr.21: 36754404-36754404 on Build GRCh38
rs377279380 Chr.21: 36754452-36754452 on Build GRCh38
rs78671947 Chr.21: 36754436-36754436 on Build GRCh38
rs780500206 Chr.21: 36754386-36754386 on Build GRCh38
rs750434336 Chr.21: 36754446-36754446 on Build GRCh38
rs1001486153 Chr.21: 36754515-36754515 on Build GRCh38
rs1046358881 Chr.21: 36754479-36754479 on Build GRCh38
rs376898721 Chr.21: 36754392-36754392 on Build GRCh38
rs778820827 Chr.21: 36754482-36754482 on Build GRCh38
rs8133818 Chr.21: 36754463-36754463 on Build GRCh38
rs149733264 Chr.21: 36754358-36754358 on Build GRCh38
rs565690761 Chr.21: 36754555-36754555 on Build GRCh38
rs775688363 Chr.21: 36754411-36754411 on Build GRCh38
rs773191073 Chr.21: 36754364-36754365 on Build GRCh38
rs777696838 Chr.21: 36754538-36754538 on Build GRCh38
rs1029184693 Chr.21: 36754529-36754529 on Build GRCh38
rs199557412 Chr.21: 36754440-36754440 on Build GRCh38
rs766703638 Chr.21: 36754513-36754513 on Build GRCh38
rs746214248 Chr.21: 36754407-36754407 on Build GRCh38
rs1056463672 Chr.21: 36754461-36754461 on Build GRCh38
rs552163792 Chr.21: 36754547-36754547 on Build GRCh38
rs780930295 Chr.21: 36754464-36754464 on Build GRCh38
rs779439814 Chr.21: 36754405-36754405 on Build GRCh38
rs935549376 Chr.21: 36754505-36754505 on Build GRCh38
rs373567395 Chr.21: 36754371-36754371 on Build GRCh38
rs556643181 Chr.21: 36754535-36754535 on Build GRCh38
rs779826873 Chr.21: 36754369-36754369 on Build GRCh38
rs761980697 Chr.21: 36754439-36754439 on Build GRCh38
rs199936080 Chr.21: 36754363-36754363 on Build GRCh38
rs373177254 Chr.21: 36754425-36754425 on Build GRCh38
rs772486641 Chr.21: 36754408-36754408 on Build GRCh38
rs762828296 Chr.21: 36754448-36754448 on Build GRCh38
rs761596791 Chr.21: 36754357-36754357 on Build GRCh38
rs148868421 Chr.21: 36754403-36754403 on Build GRCh38
rs376942757 Chr.21: 36754432-36754432 on Build GRCh38
rs538633059 Chr.21: 36754511-36754511 on Build GRCh38
rs751371980 Chr.21: 36754454-36754454 on Build GRCh38
rs139697458 Chr.21: 36754356-36754356 on Build GRCh38
rs754503384 Chr.21: 36754385-36754385 on Build GRCh38
rs145648338 Chr.21: 36754368-36754368 on Build GRCh38

View Full Product Details