Gene Transcripts

Gene Symbol
Spin4
Entrez Gene ID
270624
Gene Name
spindlin family, member 4
Gene Aliases
9630042H07Rik
Chromosome Location
Chr. X: 95022507 - 95026682 on Build GRCm38
UniGene ID
Mm.115205
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178753.4 NP_848868.1 1 4103
GenBank mRNA AK036158.1 - 1 4112

View Full Product Details