Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001077529.2 | NP_001070997.1 | 2 | 321 | |
NM_008705.5 | NP_032731.1 | 3 | 593 | ||
GenBank mRNA | AK012447.1 | - | 3 | 250 | |
AK150419.1 | - | 3 | 255 | ||
AK151262.1 | - | 2 | 210 | ||
AK169120.1 | - | 3 | 215 | ||
AV458476.1 | - | 2 | 321 | ||
BC066995.1 | - | 3 | 593 | 5691909 | |
BC086892.1 | - | 2 | 226 | 6752803 | |
BC086893.1 | - | 3 | 218 | 6758871 | |
BY274368.1 | - | 3 | 344 | ||
EF437324.1 | - | 2 | 141 | ||
X68193.1 | - | 2 | 177 |