Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005433.3 | NP_005424.1 | 4 | 605 | |
XM_005258139.3 | XP_005258196.1 | 4 | 567 | ||
XM_017025960.1 | XP_016881449.1 | 4 | 447 | ||
GenBank mRNA | AK314063.1 | - | 4 | 549 | |
BC048960.1 | - | 4 | 601 | 5260751 | |
M15990.1 | - | 4 | 591 |