Gene Transcripts

Gene Symbol
RPS6KA5
Entrez Gene ID
9252
Gene Name
ribosomal protein S6 kinase A5
Gene Aliases
MSK1, MSPK1, RLPK
Chromosome Location
Chr. 14: 90868122 - 91060649 on Build GRCh38
UniGene ID
Hs.510225
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322227.1 NP_001309156.1 4 618
NM_001322228.1 NP_001309157.1 3 478
NM_001322229.1 NP_001309158.1 3 478
NM_001322230.1 NP_001309159.1 3 478
NM_001322231.1 NP_001309160.1 3 478
NM_001322233.1 NP_001309162.1 3 478
NM_001322234.1 NP_001309163.1 3 478
NM_001322235.1 NP_001309164.1 4 618
NM_001322236.1 NP_001309165.1 3 478
NM_001322237.1 NP_001309166.1 3 277
NM_004755.3 NP_004746.2 3 478
NM_182398.2 NP_872198.1 3 478
XM_017021786.1 XP_016877275.1 3 449
GenBank mRNA AF074393.1 - 3 428
AF080000.1 - 3 444
AF090421.1 - 3 427
AK295266.1 - 3 277
AK309925.1 - 3 465
BC017187.2 - 3 429 3139114
BG699153.1 - 3 513

View Full Product Details