Gene Transcripts
- Gene Symbol
- MTMR1
- Entrez Gene ID
- 8776
- Gene Name
- myotubularin related protein 1
- Gene Aliases
- Chromosome Location
- Chr. X: 150692997 - 150765103 on Build GRCh38
- UniGene ID
- Hs.347187
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001306144.1 |
NP_001293073.1
|
7, 8 |
832 |
|
NM_001306145.1 |
NP_001293074.1
|
6, 7 |
808 |
|
NM_003828.3 |
NP_003819.1
|
6, 7 |
808 |
|
XM_005274765.3 |
XP_005274822.1
|
9, 10 |
872 |
|
XM_005274766.3 |
XP_005274823.1
|
8, 9 |
820 |
|
XM_005274767.3 |
XP_005274824.1
|
7, 8 |
839 |
|
XM_006724855.3 |
XP_006724918.1
|
6, 7 |
808 |
|
XM_011531209.2 |
XP_011529511.1
|
6, 7 |
568 |
|
XM_011531210.2 |
XP_011529512.1
|
9, 10 |
874 |
|
XM_017029920.1 |
XP_016885409.1
|
9, 10 |
872 |
|
XM_017029921.1 |
XP_016885410.1
|
7, 8 |
603 |
|
XM_017029922.1 |
XP_016885411.1
|
7, 8 |
615 |
|
XM_017029923.1 |
XP_016885412.1
|
4, 5 |
395 |
|
XM_017029924.1 |
XP_016885413.1
|
7, 8 |
839 |
|
GenBank mRNA |
AF057354.1 |
- |
6, 7 |
632 |
|
AJ224979.1 |
- |
6, 7 |
634 |
|
AK001816.1 |
- |
4, 5 |
337 |
|
AK226109.1 |
- |
7, 8 |
786 |
|
AK295738.1 |
- |
7, 8 |
627 |
|
BC032947.1 |
- |
6, 7 |
805 |
4837744 |
U58032.1 |
- |
3, 4 |
413 |
|
View Full Product Details