Gene Transcripts

Gene Symbol
PPP2R2C
Entrez Gene ID
5522
Gene Name
protein phosphatase 2 regulatory subunit Bgamma
Gene Aliases
B55-GAMMA, IMYPNO, IMYPNO1, PR52, PR55G
Chromosome Location
Chr. 4: 6320578 - 6563600 on Build GRCh38
UniGene ID
Hs.479069
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206994.1 NP_001193923.1 10 1409
NM_001206995.1 NP_001193924.1 10 1513
NM_001206996.1 NP_001193925.1 10 1563
NM_020416.3 NP_065149.2 9 1701
NM_181876.2 NP_870991.1 9 1355
XM_005247979.4 XP_005248036.1 5 1546
XM_011513495.1 XP_011511797.1 9 1450
XM_011513496.1 XP_011511798.1 9 1465
GenBank mRNA AF086924.1 - 9 1385
AF145027.1 - 9 1548
AK093115.1 - 9 1355
AK122849.1 - 5 3362
AK296474.1 - 10 1409
AK316003.1 - 11 1523
BC032954.2 - 10 1563 4823280

View Full Product Details