Gene Transcripts

Gene Symbol
USP3
Entrez Gene ID
9960
Gene Name
ubiquitin specific peptidase 3
Gene Aliases
SIH003, UBP
Chromosome Location
Chr. 15: 63504511 - 63594640 on Build GRCh38
UniGene ID
Hs.458499
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256702.1 NP_001243631.1 7 799
NM_006537.3 NP_006528.2 8 931
NR_046341.1 9 1084
NR_046342.1 9 1047
XM_017022763.1 XP_016878252.1 10 1930
XM_017022764.1 XP_016878253.1 9 1866
XM_017022765.1 XP_016878254.1 9 1072
XM_017022766.1 XP_016878255.1 10 1115
XM_017022767.1 XP_016878256.1 11 2046
XM_017022768.1 XP_016878257.1 11 2120
GenBank mRNA AF073344.1 - 8 795
AF077040.1 - 7 575
AK022931.1 - 6 518
AK300948.1 - 6 589
AK301236.1 - 7 715
AK301563.1 - 2 230
AK307810.1 - 8 690
AK309382.1 - 7 752
AK314223.1 - 8 850
AY461579.1 - 9 960
BC018113.2 - 8 796 3916811
BC065300.1 - 8 831 6146202
BC100029.1 - 8 803
BC107137.1 - 8 791 40025968
BC107138.1 - 8 791 40025969
BT007269.1 - 8 702
BX647760.1 - 9 825

View Full Product Details