Gene Transcripts
- Gene Symbol
- INPP1
- Entrez Gene ID
- 3628
- Gene Name
- inositol polyphosphate-1-phosphatase
- Gene Aliases
- Chromosome Location
- Chr. 2: 190343470 - 190371665 on Build GRCh38
- UniGene ID
- Hs.32309
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001128928.1 |
NP_001122400.1
|
3, 4 |
906 |
|
NM_002194.3 |
NP_002185.1
|
2, 3 |
762 |
|
XM_005246532.1 |
XP_005246589.1
|
2, 3 |
543 |
|
GenBank mRNA |
AB451298.1 |
- |
1, 2 |
206 |
|
AK093560.1 |
- |
4, 5 |
864 |
|
BC015496.1 |
- |
1, 2 |
272 |
3845956 |
BC106006.1 |
- |
2, 3 |
306 |
4472604 |
CR456840.1 |
- |
1, 2 |
206 |
|
CR541707.1 |
- |
1, 2 |
206 |
|
KJ535023.1 |
- |
1, 2 |
206 |
|
KJ535042.1 |
- |
1, 2 |
206 |
|
KU178146.1 |
- |
1, 2 |
206 |
|
KU178147.1 |
- |
1, 2 |
206 |
|
L08488.1 |
- |
2, 3 |
532 |
|
View Full Product Details