Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012425.3 | NP_036557.1 | 9 | 2914 | |
NM_152724.2 | NP_689937.2 | 8 | 2802 | ||
XM_005252552.3 | XP_005252609.1 | 8 | 2864 | ||
GenBank mRNA | AK055596.1 | - | 1 | 1424 | |
BX647733.1 | - | 8 | 2777 |