Gene Transcripts
- Gene Symbol
- NOTCH2
- Entrez Gene ID
- 4853
- Gene Name
- notch 2
- Gene Aliases
- AGS2, HJCYS, hN2
- Chromosome Location
- Chr. 1: 119911553 - 120073449 on Build GRCh38
- UniGene ID
- Hs.487360
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001200001.1 |
NP_001186930.1
|
13, 14 |
2514 |
|
NM_024408.3 |
NP_077719.2
|
13, 14 |
2514 |
|
XM_005270901.3 |
XP_005270958.1
|
14, 15 |
4828 |
|
XM_011541519.2 |
XP_011539821.1
|
13, 14 |
7622 |
|
XM_011541520.2 |
XP_011539822.1
|
13, 14 |
4532 |
|
XM_017001372.1 |
XP_016856861.1
|
13, 14 |
2184 |
|
XM_017001373.1 |
XP_016856862.1
|
13, 14 |
2163 |
|
GenBank mRNA |
AF308601.1 |
- |
13, 14 |
2229 |
|
AF315356.1 |
- |
13, 14 |
2229 |
|
AK307843.1 |
- |
12, 13 |
2355 |
|
BC071562.1 |
- |
13, 14 |
2463 |
4822955 |
GQ231534.1 |
- |
13, 14 |
2376 |
|
View Full Product Details