Gene Transcripts
- Gene Symbol
- SULT2A1
- Entrez Gene ID
- 6822
- Gene Name
- sulfotransferase family 2A member 1
- Gene Aliases
- DHEA-ST, DHEAS, HST, ST2, ST2A1, ST2A3, STD, hSTa
- Chromosome Location
- Chr. 19: 47870466 - 47886397 on Build GRCh38
- UniGene ID
- Hs.515835
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003167.3 |
NP_003158.2
|
2, 3 |
493 |
|
GenBank mRNA |
AK289380.1 |
- |
2, 3 |
435 |
|
AK313415.1 |
- |
2, 3 |
420 |
|
BC020755.1 |
- |
2, 3 |
410 |
4733628 |
BG563083.1 |
- |
2, 3 |
473 |
|
BX509490.1 |
- |
2, 3 |
494 |
|
L02337.1 |
- |
2, 3 |
418 |
|
L20000.1 |
- |
2, 3 |
405 |
|
S43859.1 |
- |
2, 3 |
418 |
|
S43861.1 |
- |
1, 2 |
187 |
|
U08024.1 |
- |
2, 3 |
373 |
|
U08025.1 |
- |
2, 3 |
417 |
|
X70222.1 |
- |
2, 3 |
405 |
|
X84816.1 |
- |
2, 3 |
398 |
|
View Full Product Details