Gene Transcripts

Gene Symbol
TPST2
Entrez Gene ID
8459
Gene Name
tyrosylprotein sulfotransferase 2
Gene Aliases
TANGO13B
Chromosome Location
Chr. 22: 26524082 - 26590125 on Build GRCh38
UniGene ID
Hs.632768
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008566.1 NP_001008566.1 3 523
NM_003595.3 NP_003586.3 3 415
XM_006724338.3 XP_006724401.1 4 534
GenBank mRNA AF049891.1 - 3 379
AF061254.1 - 1 187
AJ006198.1 - 1 269
AK074538.1 - 3 415
AK075139.1 - 3 487
AL136623.2 - 3 399
BC001057.2 - 3 394 3509627
BC017509.1 - 3 379 4857366
CR456597.1 - 1 277
CR533528.1 - 1 182

View Full Product Details