Gene Transcripts

Gene Symbol
LILRA2
Entrez Gene ID
11027
Gene Name
leukocyte immunoglobulin like receptor A2
Gene Aliases
CD85H, ILT1, LIR-7, LIR7
Chromosome Location
Chr. 19: 54572997 - 54587560 on Build GRCh38
UniGene ID
Hs.655593
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130917.2 NP_001124389.2 8 1585
NM_001290270.1 NP_001277199.1 6 1498
NM_001290271.1 NP_001277200.1 9 2232
NM_006866.3 NP_006857.2 8 2106
XM_006722986.1 XP_006723049.1 9 2365
XM_011526385.1 XP_011524687.1 9 2157
XM_011526386.1 XP_011524688.1 8 2329
XM_011526387.1 XP_011524689.1 7 2070
XM_011526389.1 XP_011524691.1 7 2047
XM_017026224.1 XP_016881713.1 8 2068
GenBank mRNA AF025531.1 - 8 1576
AK130170.1 - 7 1562
AK222651.1 - 7 1530
BC017412.1 - 7 1536 4618777
BC027916.1 - 7 1519 5223154
CB046242.1 - 3 492
U82275.1 - 7 1573

View Full Product Details