Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001111298.2 | NP_001104768.2 | 5 | 1148 | |
NM_001286360.1 | NP_001273289.1 | 4 | 1052 | ||
NM_001286361.1 | NP_001273290.1 | 4 | 1052 | ||
NM_173672.4 | NP_775943.1 | 5 | 1148 | ||
NR_104429.1 | 5 | 1148 | |||
XM_011535765.2 | XP_011534067.1 | 4 | 512 | ||
XM_011535766.2 | XP_011534068.1 | 5 | 616 | ||
XM_011535767.2 | XP_011534069.1 | 5 | 616 | ||
XM_011535769.2 | XP_011534071.1 | 5 | 616 | ||
XM_017010774.1 | XP_016866263.1 | 5 | 616 | ||
GenBank mRNA | AI052546.1 | - | 4 | 409 | |
AK293674.1 | - | 5 | 600 | ||
AK315790.1 | - | 5 | 621 | ||
BC038716.1 | - | 5 | 607 | 5268756 | |
BI824055.1 | - | 4 | 385 | ||
DA413621.1 | - | 4 | 435 | ||
DQ363562.1 | - | 5 | 1148 | ||
DQ423529.1 | - | 5 | 1148 |