Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017414.3 | NP_059110.2 | 11 | 1481 | |
XM_006724074.2 | XP_006724137.1 | 11 | 1552 | ||
GenBank mRNA | AF176642.2 | - | 10 | 1163 | |
AJ243526.1 | - | 11 | 1346 | ||
AL136690.1 | - | 11 | 1302 | ||
BC014896.1 | - | 11 | 1287 | 3910720 | |
CT841507.1 | - | 10 | 1209 |