Gene Transcripts
- Gene Symbol
- BRF1
- Entrez Gene ID
- 2972
- Gene Name
- BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit
- Gene Aliases
- BRF, BRF-1, CFDS, GTF3B, HEL-S-76p, TAF3B2, TAF3C, TAFIII90, TF3B90, TFIIIB90, hBRF
- Chromosome Location
- Chr. 14: 105209286 - 105315577 on Build GRCh38
- UniGene ID
- Hs.424484
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001242786.1 |
NP_001229715.1
|
5, 6 |
626 |
|
NM_001242787.1 |
NP_001229716.1
|
5, 6 |
626 |
|
NM_001242788.1 |
NP_001229717.1
|
4, 5 |
836 |
|
NM_001519.3 |
NP_001510.2
|
5, 6 |
917 |
|
NM_145685.2 |
NP_663718.1
|
1, 2 |
825 |
|
XM_005267561.3 |
XP_005267618.1
|
5, 6 |
939 |
|
XM_005267563.3 |
XP_005267620.1
|
1, 2 |
1629 |
|
XM_011536672.2 |
XP_011534974.1
|
5, 6 |
939 |
|
XM_011536674.1 |
XP_011534976.1
|
1, 2 |
1207 |
|
GenBank mRNA |
AJ297406.1 |
- |
1, 2 |
1379 |
|
AJ297407.1 |
- |
1, 2 |
833 |
|
AK294899.1 |
- |
5, 6 |
626 |
|
AK295579.1 |
- |
5, 6 |
539 |
|
BC086856.1 |
- |
4, 5 |
569 |
6166751 |
BC103859.1 |
- |
1, 2 |
338 |
|
BC103861.2 |
- |
1, 2 |
133 |
|
BC133655.1 |
- |
1, 2 |
133 |
|
U28838.1 |
- |
5, 6 |
920 |
|
U75276.1 |
- |
5, 6 |
621 |
|
View Full Product Details