Gene Transcripts

Gene Symbol
POU2F1
Entrez Gene ID
5451
Gene Name
POU class 2 homeobox 1
Gene Aliases
OCT1, OTF1, oct-1B
Chromosome Location
Chr. 1: 167220829 - 167427345 on Build GRCh38
UniGene ID
Hs.283402
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198783.1 NP_001185712.1 12 1624
NM_001198786.1 NP_001185715.1 11 1302
NM_002697.3 NP_002688.3 12 1491
NR_037163.1 13 1535
XM_011509653.2 XP_011507955.1 13 1443
XM_011509654.2 XP_011507956.1 12 1404
XM_011509655.1 XP_011507957.1 13 1443
GenBank mRNA AK091438.1 - 14 1919
AK303201.1 - 11 1240
AK308004.1 - 13 1531
AY113189.1 - 12 1443
BC001664.2 - 12 1407 2966289
BC003571.1 - 12 1407 3544378
BC007388.2 - 8 1077
BC052274.1 - 13 1515 4622256
KT438684.1 - 14 1602
S66902.1 - 12
X13403.1 - 12 1412

View Full Product Details