Gene Transcripts
- Gene Symbol
- MCC
- Entrez Gene ID
- 4163
- Gene Name
- mutated in colorectal cancers
- Gene Aliases
- MCC1
- Chromosome Location
- Chr. 5: 113022099 - 113488830 on Build GRCh38
- UniGene ID
- Hs.593171
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001085377.1 |
NP_001078846.1
|
5, 6 |
1308 |
|
NM_002387.2 |
NP_002378.1
|
3, 4 |
852 |
|
XM_005271991.2 |
XP_005272048.1
|
3, 4 |
886 |
|
XM_017009473.1 |
XP_016864962.1
|
5, 6 |
907 |
|
XM_017009474.1 |
XP_016864963.1
|
3, 4 |
3572 |
|
GenBank mRNA |
AK096212.1 |
- |
2, 3 |
454 |
|
AK128596.1 |
- |
5, 6 |
1320 |
|
AK300259.1 |
- |
3, 4 |
875 |
|
BC009279.2 |
- |
3, 4 |
852 |
4111706 |
BC018919.2 |
- |
3, 4 |
852 |
4131685 |
BX537952.1 |
- |
3, 4 |
323 |
|
BX647710.1 |
- |
4, 5 |
626 |
|
M62397.1 |
- |
3, 4 |
542 |
|
View Full Product Details