Gene Transcripts

Gene Symbol
NFYC
Entrez Gene ID
4802
Gene Name
nuclear transcription factor Y subunit gamma
Gene Aliases
CBF-C, CBFC, H1TF2A, HAP5, HSM, NF-YC
Chromosome Location
Chr. 1: 40691570 - 40771603 on Build GRCh38
UniGene ID
Hs.713051
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142587.1 NP_001136059.1 8, 9 1129
NM_001142589.1 NP_001136061.1 7, 8 1015
NM_001142590.1 NP_001136062.1 13, 14 1404
NM_001308115.1 NP_001295044.1 8, 9 1125
NM_014223.4 NP_055038.2 8, 9 1129
XM_006710658.2 XP_006710721.2 8, 9 981
XM_006710661.1 XP_006710724.1 8, 9 1010
XM_006710662.2 XP_006710725.2 8, 9 981
XM_017001364.1 XP_016856853.1 8, 9 1150
XM_017001366.1 XP_016856855.1 8, 9 1084
XM_017001368.1 XP_016856857.1 8, 9 1125
XM_017001369.1 XP_016856858.1 9, 10 1482
GenBank mRNA AB208975.1 - 8, 9 956
AK000346.1 - 8, 9 1082
AK127677.1 - 13, 14 1399
AK301385.1 - 7, 8 899
BC005003.1 - 8, 9 978 3502903
BT020081.1 - 7, 8 823
D85425.1 - 8, 9 906
D89986.1 - 8, 9 891
U62296.1 - 7, 8 823
U78774.1 - 9, 10 997
Z74792.1 - 9, 10 1008

View Full Product Details