Gene Transcripts

Gene Symbol
TRIM2
Entrez Gene ID
23321
Gene Name
tripartite motif containing 2
Gene Aliases
CMT2R, RNF86
Chromosome Location
Chr. 4: 153152281 - 153339322 on Build GRCh38
UniGene ID
Hs.435711
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130067.1 NP_001123539.1 2 294
NM_001302692.1 NP_001289621.1 2 291
NM_001302693.1 NP_001289622.1 2 288
NM_001302694.1 NP_001289623.1 2 393
NM_015271.4 NP_056086.2 2 291
XM_006714157.1 XP_006714220.1 2 291
XM_006714158.1 XP_006714221.1 2 256
XM_006714159.3 XP_006714222.1 2 1102
XM_006714160.1 XP_006714223.1 3 1056
XM_006714161.1 XP_006714224.1 3 1053
XM_006714162.1 XP_006714225.1 3 773
XM_011531794.1 XP_011530096.1 2 370
XM_011531795.1 XP_011530097.1 3 776
XM_011531796.2 XP_011530098.1 2 306
XM_011531798.2 XP_011530100.1 2 411
XM_017007943.1 XP_016863432.1 2 256
XM_017007944.1 XP_016863433.1 3 1056
XM_017007945.1 XP_016863434.1 3 1053
XM_017007946.1 XP_016863435.1 2 1099
XM_017007947.1 XP_016863436.1 2 370
XM_017007948.1 XP_016863437.1 2 324
XM_017007949.1 XP_016863438.1 3 773
XM_017007950.1 XP_016863439.1 3 776
XM_017007951.1 XP_016863440.1 2 470
XM_017007952.1 XP_016863441.1 2 478
XM_017007954.1 XP_016863443.1 2 247
XM_017007955.1 XP_016863444.1 2 240
GenBank mRNA AB011089.1 - 2 239
AF220018.1 - 2 239
AK308731.1 - 2 259
BC011052.1 - 2 294 4156234
BP310511.1 - 2 392
DA310963.1 - 2 378
DA407606.1 - 2 256
DA631098.1 - 2 292

View Full Product Details