Gene Transcripts

Gene Symbol
FEV
Entrez Gene ID
54738
Gene Name
FEV, ETS transcription factor
Gene Aliases
HSRNAFEV, PET-1
Chromosome Location
Chr. 2: 218981087 - 218985657 on Build GRCh38
UniGene ID
Hs.234759
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017521.2 NP_059991.1 3 1666
GenBank mRNA BC023511.2 - 3 1561
Y08976.1 - 3 1668

View Full Product Details