Gene Transcripts

Gene Symbol
TRIM8
Entrez Gene ID
81603
Gene Name
tripartite motif containing 8
Gene Aliases
GERP, RNF27
Chromosome Location
Chr. 10: 102644484 - 102658319 on Build GRCh38
UniGene ID
Hs.336810
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030912.2 NP_112174.2 2 761
GenBank mRNA AF220034.1 - 2 638
AF281046.1 - 2 647
AL360160.1 - 2 647
BC021925.1 - 2 760 4857393

View Full Product Details