Gene Transcripts

Gene Symbol
PON1
Entrez Gene ID
5444
Gene Name
paraoxonase 1
Gene Aliases
ESA, MVCD5, PON
Chromosome Location
Chr. 7: 95298357 - 95324572 on Build GRCh38
UniGene ID
Hs.370995
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000446.5 NP_000437.3 2 211
GenBank mRNA AK314027.1 - 2 137
BC074719.2 - 2 143 30915169
BJ993711.1 - 2 211
CB122285.1 - 2 171
D84371.1 - 2 168
M63012.1 - 2 123
M63013.1 - 2 62
M63014.1 - 1 28
S64615.1 - 2 123
U53784.1 - 2 168
Z70723.1 - 2 168

View Full Product Details