Gene Transcripts

Gene Symbol
GYG2
Entrez Gene ID
8908
Gene Name
glycogenin 2
Gene Aliases
GN-2, GN2
Chromosome Location
Chr. X: 2828712 - 2882820 on Build GRCh38
UniGene ID
Hs.567381
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079855.1 NP_001073324.1 11 1857
NM_001184702.1 NP_001171631.1 11 1854
NM_001184703.1 NP_001171632.1 10 1737
NM_001184704.1 NP_001171633.1 8 1502
NM_003918.2 NP_003909.2 12 1950
XM_011545600.2 XP_011543902.1 11 1788
XM_017029927.1 XP_016885416.1 12 1900
XM_017029928.1 XP_016885417.1 12 1875
XM_017029929.1 XP_016885418.1 12 2057
XM_017029930.1 XP_016885419.1 9 1552
XM_017029931.1 XP_016885420.1 9 1575
GenBank mRNA AK098046.1 - 8 1410
AK292496.1 - 11 1765
BC023152.1 - 11 1724 3923041
U94357.1 - 2 404
U94360.1 - 6 1192
U94362.1 - 12 1951
U94363.1 - 11 1728

View Full Product Details