Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286123.1 | NP_001273052.1 | 2 | 193 | |
NM_001286125.1 | NP_001273054.1 | 2 | 193 | ||
NM_005835.3 | NP_005826.1 | 3 | 643 | ||
XM_005248784.2 | XP_005248841.1 | 3 | 643 | ||
XM_006714949.3 | XP_006715012.1 | 3 | 380 | ||
XM_006714950.2 | XP_006715013.1 | 2 | 270 | ||
XM_006714951.1 | XP_006715014.1 | 3 | 643 | ||
XM_017010159.1 | XP_016865648.1 | 2 | 533 | ||
XM_017010160.1 | XP_016865649.1 | 3 | 638 | ||
GenBank mRNA | BC104822.1 | - | 2 | 166 | |
BC112033.1 | - | 2 | 166 | 8327501 | |
BC143323.1 | - | 2 | 164 | ||
U90544.1 | - | 3 | 528 |